Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC77
Entrez Gene ID
84318
Gene Name
coiled-coil domain containing 77
Gene Aliases
-
Chromosome Location
Chr.12: 389350 - 442642 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130146.1 NP_001123618.1 2 - 3 166 86
NM_001130147.1 NP_001123619.1 2 - 3 193 86
GenBank mRNA AK293165.1 - 2 - 3 104 86
AK308216.1 - 2 - 3 148 86
CX784327.1 - 2 - 3 73 86

View Full Product Details