The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130146.1 | NP_001123618.1 | 3 - 4 | 397 | 81 |
NM_001130147.1 | NP_001123619.1 | 3 - 4 | 424 | 81 | |
NM_001130148.1 | NP_001123620.1 | 2 - 3 | 370 | 81 | |
NM_032358.3 | NP_115734.1 | 4 - 5 | 451 | 81 | |
GenBank mRNA | AK027638.1 | - | 4 - 5 | 369 | 81 |
AK293165.1 | - | 3 - 4 | 335 | 81 | |
AK308216.1 | - | 3 - 4 | 379 | 81 | |
AK316029.1 | - | 2 - 3 | 372 | 81 | |
BC006444.2 | - | 3 - 4 | 340 | 81 | |
CX784327.1 | - | 3 - 4 | 304 | 81 | |
DA711064.1 | - | 4 - 5 | 452 | 81 | |
DC331633.1 | - | 2 - 3 | 370 | 81 |