The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130146.1 | NP_001123618.1 | 6 - 7 | 713 | 123 |
NM_001130147.1 | NP_001123619.1 | 6 - 7 | 740 | 123 | |
NM_001130148.1 | NP_001123620.1 | 5 - 6 | 686 | 123 | |
NM_032358.3 | NP_115734.1 | 7 - 8 | 767 | 123 | |
GenBank mRNA | AK027638.1 | - | 7 - 8 | 685 | 123 |
AK293165.1 | - | 6 - 7 | 651 | 123 | |
AK308216.1 | - | 6 - 7 | 695 | 123 | |
AK316029.1 | - | 5 - 6 | 688 | 123 | |
BC006444.2 | - | 6 - 7 | 656 | 123 | |
CX784327.1 | - | 6 - 7 | 620 | 123 |