Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXL13
Entrez Gene ID
222235
Gene Name
F-box and leucine rich repeat protein 13
Gene Aliases
DRC6, Fbl13
Chromosome Location
Chr.7: 102699226 - 103074841 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111038.1 NP_001104508.1 5 - 6 573 111
NM_001287150.1 NP_001274079.1 5 - 6 573 111
NM_145032.3 NP_659469.3 5 - 6 750 111
NR_105043.1 6 - 7 664 111
XM_005250205.3 XP_005250262.1 6 - 7 656 111
XM_005250207.3 XP_005250264.1 6 - 7 656 111
XM_005250208.3 XP_005250265.1 6 - 7 656 111
XM_005250209.2 XP_005250266.1 5 - 6 554 111
XM_011515928.2 XP_011514230.1 6 - 7 656 111
XM_011515929.2 XP_011514231.1 6 - 7 656 111
XM_011515930.2 XP_011514232.1 5 - 6 620 111
XM_011515932.2 XP_011514234.1 6 - 7 656 111
XM_017011850.1 XP_016867339.1 6 - 7 600 111
XM_017011851.1 XP_016867340.1 6 - 7 656 111
XM_017011852.1 XP_016867341.1 6 - 7 806 111
XM_017011853.1 XP_016867342.1 5 - 6 606 111
GenBank mRNA AK097537.1 - 5 - 6 750 111
BC026121.1 - 6 - 7 667 111
BC031285.1 - 5 - 6 565 111

View Full Product Details