Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTOA
Entrez Gene ID
146183
Gene Name
otoancorin
Gene Aliases
CT108, DFNB22
Chromosome Location
Chr.16: 21663857 - 21771468 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161683.1 NP_001155155.1 10 - 11 1484 93
NM_144672.3 NP_653273.3 14 - 15 1628 93
NM_170664.2 NP_733764.1 5 - 6 722 93
XM_011545747.2 XP_011544049.1 15 - 16 2007 93
XM_011545748.2 XP_011544050.2 4 - 5 1001 93
XM_017022950.1 XP_016878439.1 15 - 16 1731 93
GenBank mRNA AK093062.1 - 5 - 6 722 93
AK125840.1 - 10 - 11 1484 93
AK302733.1 - 4 - 5 784 93
BC129992.1 - 14 - 15 1625 93
BC129993.1 - 14 - 15 1632 93

View Full Product Details