The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172435.1 | NP_001165906.1 | 6 - 7 | 522 | 88 |
NM_012233.2 | NP_036365.1 | 6 - 7 | 522 | 88 | |
XM_011510823.2 | XP_011509125.1 | 6 - 7 | 522 | 88 | |
XM_011510825.2 | XP_011509127.1 | 6 - 7 | 522 | 88 | |
GenBank mRNA | AB449877.1 | - | 6 - 7 | 479 | 88 |
BC146809.1 | - | 8 - 9 | 759 | 88 | |
BP872264.1 | - | 6 - 7 | 503 | 88 | |
D31886.1 | - | 6 - 7 | 481 | 88 | |
DA333844.1 | - | 6 - 7 | 522 | 88 |