The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172435.1 | NP_001165906.1 | 9 - 10 | 871 | 66 |
NM_012233.2 | NP_036365.1 | 9 - 10 | 871 | 66 | |
XM_011510823.2 | XP_011509125.1 | 9 - 10 | 871 | 66 | |
XM_011510825.2 | XP_011509127.1 | 9 - 10 | 871 | 66 | |
GenBank mRNA | AB449877.1 | - | 9 - 10 | 828 | 66 |
AK022494.1 | - | 9 - 10 | 843 | 66 | |
BC146809.1 | - | 11 - 12 | 1108 | 66 | |
D31886.1 | - | 9 - 10 | 830 | 66 |