Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2orf72
Entrez Gene ID
257407
Gene Name
chromosome 2 open reading frame 72
Gene Aliases
-
Chromosome Location
Chr.2: 231037566 - 231049714 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144994.1 NP_001138466.1 1 - 2 630 151

View Full Product Details