Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP13A1
Entrez Gene ID
57130
Gene Name
ATPase 13A1
Gene Aliases
ATP13A, CGI-152
Chromosome Location
Chr.19: 19645198 - 19663694 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020410.2 NP_065143.2 18 - 19 2564 86
GenBank mRNA AB058728.1 - 18 - 19 2534 86
AF288687.1 - 18 - 19 2182 86
AK026044.1 - 2 - 3 714 86
AK056420.1 - 18 - 19 2540 86
AK074159.1 - 10 - 11 3279 86
AK074798.1 - 7 - 8 999 86
AK074920.1 - 7 - 8 1144 86
AK123780.1 - 1 - 2 908 86
AK125861.1 - 2 - 3 2861 86
AK130125.1 - 9 - 10 1204 86
AK172778.1 - 17 - 18 1995 86

View Full Product Details