The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020410.2 | NP_065143.2 | 5 - 6 | 940 | 56 |
GenBank mRNA | AB058728.1 | - | 5 - 6 | 910 | 56 |
AF288687.1 | - | 5 - 6 | 558 | 56 | |
AK056420.1 | - | 5 - 6 | 916 | 56 | |
AK172778.1 | - | 3 - 4 | 371 | 56 |