The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143824.1 | NP_001137296.1 | 6 - 7 | 701 | 127 |
NM_018018.4 | NP_060488.2 | 7 - 8 | 893 | 127 | |
XM_005268997.2 | XP_005269054.1 | 6 - 7 | 702 | 127 | |
GenBank mRNA | AB055003.1 | - | 5 - 6 | 525 | 127 |
AF193836.1 | - | 6 - 7 | 681 | 127 | |
AF305814.1 | - | 7 - 8 | 858 | 127 | |
AK001053.1 | - | 7 - 8 | 769 | 127 | |
AK291168.1 | - | 6 - 7 | 702 | 127 | |
BC069819.1 | - | 6 - 7 | 754 | 127 | |
BC101827.1 | - | 5 - 6 | 538 | 127 | |
BC104913.1 | - | 5 - 6 | 537 | 127 |