Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ESRP2
Entrez Gene ID
80004
Gene Name
epithelial splicing regulatory protein 2
Gene Aliases
RBM35B
Chromosome Location
Chr.16: 68228547 - 68236233 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024939.2 NP_079215.2 13 - 14 2079 80
XM_005256153.4 XP_005256210.1 13 - 14 2018 80
XM_006721273.3 XP_006721336.1 13 - 14 2087 80
XM_011523338.2 XP_011521640.1 12 - 13 1979 80
GenBank mRNA AF289577.1 - 7 - 8 1181 80
AK025571.1 - 13 - 14 2008 80
AK025901.1 - 2 - 3 344 80
BC030146.2 - 13 - 14 1901 80
BC069241.1 - 13 - 14 2430 80

View Full Product Details