Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ESRP2
Entrez Gene ID
80004
Gene Name
epithelial splicing regulatory protein 2
Gene Aliases
RBM35B
Chromosome Location
Chr.16: 68228547 - 68236233 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024939.2 NP_079215.2 1 - 2 388 70
XM_005256153.4 XP_005256210.1 1 - 2 330 70
GenBank mRNA AK025571.1 - 1 - 2 287 70
BC030146.2 - 1 - 2 210 70
BC069241.1 - 1 - 2 739 70
BU943912.1 - 1 - 2 222 70
LN680438.1 - 1 - 2 200 70
LN680439.1 - 1 - 2 200 70

View Full Product Details