The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143824.1 | NP_001137296.1 | 11 - 12 | 1287 | 80 |
NM_018018.4 | NP_060488.2 | 12 - 13 | 1479 | 80 | |
XM_005268997.2 | XP_005269054.1 | 11 - 12 | 1288 | 80 | |
GenBank mRNA | AB055003.1 | - | 10 - 11 | 1111 | 80 |
AF193836.1 | - | 11 - 12 | 1267 | 80 | |
AF305814.1 | - | 12 - 13 | 1444 | 80 | |
AK291168.1 | - | 11 - 12 | 1288 | 80 | |
BC069819.1 | - | 11 - 12 | 1340 | 80 | |
BC101827.1 | - | 10 - 11 | 1124 | 80 | |
BC104913.1 | - | 10 - 11 | 1123 | 80 |