The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142588.1 | NP_001136060.1 | 8 - 9 | 1128 | 73 |
NM_001308114.1 | NP_001295043.1 | 7 - 8 | 830 | 73 | |
XM_005270894.1 | XP_005270951.1 | 8 - 9 | 1124 | 73 | |
XM_005270895.3 | XP_005270952.3 | 8 - 9 | 980 | 73 | |
XM_006710660.2 | XP_006710723.2 | 8 - 9 | 980 | 73 | |
XM_011541516.1 | XP_011539818.1 | 8 - 9 | 929 | 73 | |
XM_011541517.1 | XP_011539819.1 | 8 - 9 | 980 | 73 | |
XM_017001365.1 | XP_016856854.1 | 8 - 9 | 1009 | 73 | |
XM_017001367.1 | XP_016856856.1 | 8 - 9 | 1124 | 73 | |
GenBank mRNA | AF191744.1 | - | 7 - 8 | 829 | 73 |
AK300774.1 | - | 8 - 9 | 931 | 73 |