The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145207.2 | NP_660208.2 | 14 - 15 | 2417 | 84 |
XM_005262783.4 | XP_005262840.1 | 14 - 15 | 2414 | 84 | |
XM_011531678.2 | XP_011529980.1 | 15 - 16 | 2486 | 84 | |
XM_017007825.1 | XP_016863314.1 | 15 - 16 | 2485 | 84 | |
XM_017007826.1 | XP_016863315.1 | 15 - 16 | 2485 | 84 | |
XM_017007828.1 | XP_016863317.1 | 14 - 15 | 2379 | 84 | |
XM_017007829.1 | XP_016863318.1 | 13 - 14 | 3109 | 84 | |
GenBank mRNA | AF361489.1 | - | 14 - 15 | 2413 | 84 |
AK091384.1 | - | 14 - 15 | 2372 | 84 | |
AL832175.1 | - | 14 - 15 | 2410 | 84 |