The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017446.3 | NP_059142.2 | 3 - 4 | 434 | 127 |
NM_080794.3 | NP_542984.2 | 3 - 4 | 434 | 127 | |
XM_006724026.3 | XP_006724089.1 | 3 - 4 | 426 | 127 | |
XM_011529651.2 | XP_011527953.1 | 3 - 4 | 505 | 127 | |
GenBank mRNA | AF109357.1 | - | 3 - 4 | 408 | 127 |
BC004896.2 | - | 3 - 4 | 407 | 127 | |
BC107719.1 | - | 3 - 4 | 430 | 127 | |
BQ719968.1 | - | 3 - 4 | 462 | 127 |