The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261392.1 | NP_001248321.1 | 9 - 10 | 805 | 69 |
NM_024894.3 | NP_079170.2 | 10 - 11 | 883 | 69 | |
NR_048552.1 | 9 - 10 | 837 | 69 | ||
XM_011510398.2 | XP_011508700.1 | 9 - 10 | 750 | 69 | |
XM_011510399.2 | XP_011508701.1 | 8 - 9 | 688 | 69 | |
XM_011510400.2 | XP_011508702.1 | 10 - 11 | 871 | 69 | |
XM_011510402.2 | XP_011508704.1 | 10 - 11 | 871 | 69 | |
GenBank mRNA | AK024000.1 | - | 9 - 10 | 724 | 69 |
AK290680.1 | - | 10 - 11 | 847 | 69 | |
AK297169.1 | - | 9 - 10 | 793 | 69 | |
AK299617.1 | - | 9 - 10 | 825 | 69 | |
BC005125.2 | - | 10 - 11 | 863 | 69 | |
BG390065.1 | - | 5 - 6 | 337 | 69 |