The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006137.6 | NP_006128.1 | 1 - 2 | 192 | 125 |
GenBank mRNA | AK223465.1 | - | 1 - 2 | 164 | 125 |
AK298096.1 | - | 1 - 2 | 117 | 125 | |
AK310061.1 | - | 1 - 2 | 119 | 125 | |
AK310558.1 | - | 1 - 2 | 165 | 125 | |
AY935534.1 | - | 1 - 2 | 164 | 125 | |
AY935535.1 | - | 1 - 2 | 164 | 125 | |
AY935536.1 | - | 1 - 2 | 164 | 125 | |
AY935537.1 | - | 1 - 2 | 164 | 125 | |
BC009293.2 | - | 1 - 2 | 136 | 125 | |
BC013297.2 | - | 1 - 2 | 136 | 125 | |
BI838777.1 | - | 1 - 2 | 193 | 125 | |
BM917451.1 | - | - | 174 | 125 | |
BT006696.1 | - | 1 - 2 | 83 | 125 |