Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMYM3
Entrez Gene ID
9203
Gene Name
zinc finger MYM-type containing 3
Gene Aliases
DXS6673E, MYM, XFIM, ZNF198L2, ZNF261
Chromosome Location
Chr.X: 71239624 - 71255261 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171162.1 NP_001164633.1 23 - 24 4516 86
NM_005096.3 NP_005087.1 23 - 24 3991 86
NM_201599.2 NP_963893.1 23 - 24 3982 86
XM_005262309.3 XP_005262366.1 23 - 24 4425 86
XM_005262310.2 XP_005262367.1 23 - 24 3902 86
XM_011531062.2 XP_011529364.1 23 - 24 3891 86
GenBank mRNA AB002383.1 - 23 - 24 3890 86
BC069057.1 - 23 - 24 3895 86
X95808.1 - 24 - 25 4492 86

View Full Product Details