The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171162.1 | NP_001164633.1 | 3 - 4 | 1457 | 98 |
NM_001171163.1 | NP_001164634.1 | 3 - 4 | 896 | 98 | |
NM_005096.3 | NP_005087.1 | 3 - 4 | 896 | 98 | |
NM_201599.2 | NP_963893.1 | 3 - 4 | 887 | 98 | |
XM_005262309.3 | XP_005262366.1 | 3 - 4 | 1330 | 98 | |
XM_005262310.2 | XP_005262367.1 | 3 - 4 | 843 | 98 | |
XM_011531062.2 | XP_011529364.1 | 3 - 4 | 796 | 98 | |
GenBank mRNA | AB002383.1 | - | 3 - 4 | 795 | 98 |
AK290762.1 | - | 3 - 4 | 896 | 98 | |
BC013009.2 | - | 3 - 4 | 820 | 98 | |
BC069057.1 | - | 3 - 4 | 800 | 98 | |
BT007095.1 | - | 2 - 3 | 708 | 98 | |
X95808.1 | - | 4 - 5 | 1433 | 98 |