Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANXA6
Entrez Gene ID
309
Gene Name
annexin A6
Gene Aliases
ANX6, CBP68
Chromosome Location
Chr.5: 151100706 - 151157882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001155.4 NP_001146.2 13 - 14 1203 71
NM_001193544.1 NP_001180473.1 12 - 13 1080 71
XM_005268432.4 XP_005268489.1 13 - 14 1203 71
GenBank mRNA AK126836.1 - - 1188 71
AK290672.1 - 13 - 14 1068 71
AK298582.1 - 12 - 13 860 71
AK303078.1 - 12 - 13 1066 71
BC017046.1 - 13 - 14 1024 71
D00510.1 - 12 - 13 975 71
J03578.1 - 13 - 14 1031 71
KU177903.1 - 12 - 13 975 71
KU177904.1 - 12 - 13 975 71
Y00097.1 - 13 - 14 1075 71

View Full Product Details