Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GFPT2
Entrez Gene ID
9945
Gene Name
glutamine-fructose-6-phosphate transaminase 2
Gene Aliases
GFAT, GFAT 2, GFAT2
Chromosome Location
Chr.5: 180300690 - 180353387 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005110.3 NP_005101.1 18 - 19 2181 61
XM_017010100.1 XP_016865589.1 17 - 18 2066 61
GenBank mRNA AB016789.1 - 18 - 19 2110 61
AK001242.1 - 8 - 9 1063 61
AK022507.1 - 18 - 19 2121 61
AK297750.1 - 16 - 17 1817 61
AK314551.1 - 18 - 19 2121 61
BC000012.2 - 18 - 19 2119 61
BT009818.1 - 17 - 18 2011 61
KU178494.1 - 17 - 18 2011 61

View Full Product Details