Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP13A1
Entrez Gene ID
57130
Gene Name
ATPase 13A1
Gene Aliases
ATP13A, CGI-152
Chromosome Location
Chr.19: 19645198 - 19663694 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK074159.1 - 6 - 7 877 81
AK074920.1 - 2 - 3 491 81

View Full Product Details