Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUPT4H1
Entrez Gene ID
6827
Gene Name
SPT4 homolog, DSIF elongation factor subunit
Gene Aliases
SPT4, SPT4H, SUPT4H, Supt4a
Chromosome Location
Chr.17: 58345175 - 58352238 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003168.2 NP_003159.1 2 - 3 278 108
NR_073470.1 2 - 3 280 108
GenBank mRNA AK129758.1 - 2 - 3 249 108
AK311986.1 - 2 - 3 191 108
BC002802.2 - 2 - 3 236 108
BP196741.1 - 2 - 3 278 108
BP318625.1 - 2 - 3 241 108
BX331655.2 - 2 - 3 242 108
CR407663.1 - 2 - 3 175 108
U38817.1 - 2 - 3 217 108
U38818.1 - 2 - 3 236 108
U43923.1 - 2 - 3 242 108

View Full Product Details