The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005259204.3 | XP_005259261.1 | 2 - 3 | 872 | 87 |
GenBank mRNA | AK308893.1 | - | 1 - 2 | 184 | 87 |
AK310381.1 | - | 1 - 2 | 224 | 87 |