Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BRAF
Entrez Gene ID
673
Gene Name
B-Raf proto-oncogene, serine/threonine kinase
Gene Aliases
B-RAF1, B-raf, BRAF1, NS7, RAFB1
Chromosome Location
Chr.7: 140719331 - 140924764 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004333.4 NP_004324.2 11 - 12 1494 67
XM_005250045.2 XP_005250102.1 11 - 12 1441 67
XM_017012558.1 XP_016868047.1 12 - 13 1561 67
XM_017012559.1 XP_016868048.1 12 - 13 1561 67
GenBank mRNA AK314346.1 - 11 - 12 1441 67
BC038966.1 - 1 - 2 141 67
BC101757.1 - 11 - 12 1455 67
BC112079.1 - 11 - 12 1455 67
M21001.1 - 1 - 2 119 67
M95712.2 - 11 - 12 1494 67

Gene Symbol
KIAA1549
Entrez Gene ID
57670
Gene Name
KIAA1549
Gene Aliases
-
Chromosome Location
Chr.7: 138831377 - 138981626 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AM989477.1 - - 1640 67

View Full Product Details