Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPATA5
Entrez Gene ID
166378
Gene Name
spermatogenesis associated 5
Gene Aliases
AFG2, EHLMRS, SPAF
Chromosome Location
Chr.4: 122923070 - 123319450 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145207.2 NP_660208.2 1 - 2 240 131
XM_011531679.2 XP_011529981.1 1 - 2 195 131
XM_017007825.1 XP_016863314.1 1 - 2 236 131
XM_017007826.1 XP_016863315.1 1 - 2 236 131
XM_017007827.1 XP_016863316.1 1 - 2 236 131
XM_017007830.1 XP_016863319.1 1 - 2 236 131
GenBank mRNA AF361489.1 - 1 - 2 236 131
AF479656.1 - 1 - 2 210 131
AK091384.1 - 1 - 2 195 131
AL832175.1 - 1 - 2 233 131

View Full Product Details