Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNBP1L
Entrez Gene ID
54874
Gene Name
formin binding protein 1 like
Gene Aliases
C1orf39, TOCA1
Chromosome Location
Chr.1: 93448067 - 93569671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024948.2 NP_001020119.1 8 - 9 942 102
NM_001164473.2 NP_001157945.1 8 - 9 942 102
NM_017737.4 NP_060207.2 8 - 9 942 102
XM_011541625.2 XP_011539927.1 8 - 9 1006 102
XM_017001533.1 XP_016857022.1 9 - 10 1137 102
GenBank mRNA AK000282.1 - 2 - 3 194 102
AK299760.1 - 3 - 4 328 102
AK299943.1 - 2 - 3 238 102
AY514449.1 - 8 - 9 791 102
BC062477.1 - 3 - 4 393 102
BC074891.2 - 2 - 3 172 102
BC074892.2 - 2 - 3 172 102

View Full Product Details