The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032449.2 | NP_115825.1 | 19 - 20 | 2298 | 101 |
XM_005270590.2 | XP_005270647.1 | 19 - 20 | 2384 | 101 | |
XM_005270591.2 | XP_005270648.1 | 19 - 20 | 2366 | 101 | |
XM_005270593.2 | XP_005270650.1 | 15 - 16 | 2120 | 101 | |
XM_006710423.2 | XP_006710486.1 | 18 - 19 | 2258 | 101 | |
XM_017000559.1 | XP_016856048.1 | 19 - 20 | 2296 | 101 | |
GenBank mRNA | AB058739.1 | - | 15 - 16 | 1729 | 101 |
BC007912.2 | - | 7 - 8 | 746 | 101 | |
BC014080.2 | - | 15 - 16 | 1780 | 101 | |
BC016880.1 | - | 13 - 14 | 1489 | 101 | |
BC039308.1 | - | 17 - 18 | 3122 | 101 |