The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032449.2 | NP_115825.1 | 21 - 22 | 2402 | 78 |
XM_005270590.2 | XP_005270647.1 | 21 - 22 | 2488 | 78 | |
XM_005270591.2 | XP_005270648.1 | 21 - 22 | 2470 | 78 | |
XM_005270593.2 | XP_005270650.1 | 17 - 18 | 2224 | 78 | |
XM_006710423.2 | XP_006710486.1 | 20 - 21 | 2362 | 78 | |
XM_017000559.1 | XP_016856048.1 | 21 - 22 | 2400 | 78 | |
GenBank mRNA | AB058739.1 | - | 17 - 18 | 1833 | 78 |
BC007912.2 | - | 9 - 10 | 850 | 78 | |
BC014080.2 | - | 17 - 18 | 1884 | 78 | |
BC016880.1 | - | 15 - 16 | 1593 | 78 | |
BC039308.1 | - | 19 - 20 | 3226 | 78 |