The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024576.4 | NP_078852.3 | 2 - 3 | 818 | 130 |
XM_017011304.1 | XP_016866793.1 | 2 - 3 | 380 | 130 | |
GenBank mRNA | BC018688.1 | - | 2 - 3 | 453 | 130 |
BC110999.1 | - | 2 - 3 | 457 | 130 |