The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014614.2 | NP_055429.2 | 8 - 9 | 1017 | 131 |
XM_006711969.1 | XP_006712032.1 | 6 - 7 | 533 | 131 | |
XM_011532705.2 | XP_011531007.1 | 9 - 10 | 1041 | 131 | |
XM_011532706.2 | XP_011531008.1 | 9 - 10 | 1041 | 131 | |
XM_011532707.2 | XP_011531009.1 | 8 - 9 | 975 | 131 | |
XM_011532709.2 | XP_011531011.1 | 9 - 10 | 1041 | 131 | |
GenBank mRNA | AB621805.1 | - | 7 - 8 | 1134 | 131 |
AY894754.1 | - | 8 - 9 | 1017 | 131 | |
AY894756.1 | - | 8 - 9 | 961 | 131 | |
BC112169.1 | - | 7 - 8 | 653 | 131 | |
BC113668.1 | - | 7 - 8 | 653 | 131 | |
BC143739.1 | - | 7 - 8 | 653 | 131 | |
D38521.1 | - | 8 - 9 | 827 | 131 |