The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207330.2 | NP_997213.1 | 3 - 4 | 444 | 71 |
XM_017007784.1 | XP_016863273.1 | 3 - 4 | 401 | 71 | |
GenBank mRNA | AK095299.1 | - | 3 - 4 | 403 | 71 |
BC067881.1 | - | 3 - 4 | 442 | 71 | |
CR749484.1 | - | 3 - 4 | 443 | 71 |