Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRRT
Entrez Gene ID
51593
Gene Name
serrate, RNA effector molecule
Gene Aliases
ARS2, ASR2, serrate
Chromosome Location
Chr.7: 100874971 - 100888664 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128852.1 NP_001122324.1 15 - 16 2240 71
NM_001128853.1 NP_001122325.1 15 - 16 2243 71
NM_001128854.1 NP_001122326.1 15 - 16 2240 71
NM_015908.5 NP_056992.4 15 - 16 2243 71
XM_005250405.2 XP_005250462.1 15 - 16 2100 71
XM_005250406.2 XP_005250463.1 15 - 16 2097 71
XM_005250407.2 XP_005250464.1 15 - 16 2100 71
XM_005250408.2 XP_005250465.1 15 - 16 2097 71
XM_017012290.1 XP_016867779.1 16 - 17 2425 71
XM_017012291.1 XP_016867780.1 16 - 17 2422 71
GenBank mRNA AF082871.1 - 5 - 6 651 71
AF112994.1 - 2 - 3 426 71
AF248955.1 - 15 - 16 2206 71
AK296131.1 - 9 - 10 1209 71
AK302820.1 - 15 - 16 2132 71
AL096723.1 - 15 - 16 2194 71
BC000082.2 - 6 - 7 764 71
BC069249.1 - 13 - 14 1784 71
BC109117.1 - 15 - 16 2134 71
BQ059529.1 - 3 - 4 539 71

View Full Product Details