The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128852.1 | NP_001122324.1 | 4 - 5 | 669 | 58 |
NM_001128853.1 | NP_001122325.1 | 4 - 5 | 669 | 58 | |
NM_001128854.1 | NP_001122326.1 | 4 - 5 | 669 | 58 | |
NM_015908.5 | NP_056992.4 | 4 - 5 | 669 | 58 | |
XM_005250405.2 | XP_005250462.1 | 4 - 5 | 526 | 58 | |
XM_005250406.2 | XP_005250463.1 | 4 - 5 | 526 | 58 | |
XM_005250407.2 | XP_005250464.1 | 4 - 5 | 526 | 58 | |
XM_005250408.2 | XP_005250465.1 | 4 - 5 | 526 | 58 | |
XM_017012290.1 | XP_016867779.1 | 5 - 6 | 851 | 58 | |
XM_017012291.1 | XP_016867780.1 | 5 - 6 | 851 | 58 | |
GenBank mRNA | AF248955.1 | - | 4 - 5 | 632 | 58 |
AK302820.1 | - | 4 - 5 | 669 | 58 | |
AL096723.1 | - | 4 - 5 | 624 | 58 | |
BC069249.1 | - | 2 - 3 | 210 | 58 | |
BC109117.1 | - | 4 - 5 | 563 | 58 | |
BG477711.1 | - | - | 52 | 58 |