The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024924.1 | NP_001020095.1 | 13 - 14 | 1959 | 109 |
NM_018261.3 | NP_060731.2 | 13 - 14 | 1803 | 109 | |
NM_178237.2 | NP_839955.1 | 12 - 13 | 1758 | 109 | |
XM_005265747.2 | XP_005265804.1 | 14 - 15 | 2047 | 109 | |
XM_005265748.1 | XP_005265805.1 | 14 - 15 | 1859 | 109 | |
XM_005265750.2 | XP_005265807.1 | 12 - 13 | 1835 | 109 | |
XM_017008405.1 | XP_016863894.1 | 13 - 14 | 2002 | 109 | |
XM_017008406.1 | XP_016863895.1 | 13 - 14 | 1814 | 109 | |
XM_017008408.1 | XP_016863897.1 | 6 - 7 | 2057 | 109 | |
XM_017008409.1 | XP_016863898.1 | 5 - 6 | 2012 | 109 | |
GenBank mRNA | AF208854.1 | - | 4 - 5 | 430 | 109 |
AK001755.1 | - | 13 - 14 | 1745 | 109 | |
AK023461.1 | - | 4 - 5 | 1517 | 109 | |
AK027047.1 | - | 8 - 9 | 906 | 109 | |
AK027413.1 | - | 12 - 13 | 1675 | 109 | |
AL833208.1 | - | 11 - 12 | 1546 | 109 | |
BC020650.1 | - | 4 - 5 | 427 | 109 | |
BC094751.1 | - | 13 - 14 | 1959 | 109 |