Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOC1
Entrez Gene ID
55763
Gene Name
exocyst complex component 1
Gene Aliases
BM-102, SEC3, SEC3L1, SEC3P
Chromosome Location
Chr.4: 55853616 - 55905078 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024924.1 NP_001020095.1 7 - 8 1278 65
NM_018261.3 NP_060731.2 7 - 8 1122 65
NM_178237.2 NP_839955.1 7 - 8 1122 65
XM_005265747.2 XP_005265804.1 7 - 8 1312 65
XM_005265748.1 XP_005265805.1 7 - 8 1124 65
XM_005265750.2 XP_005265807.1 7 - 8 1199 65
XM_017008405.1 XP_016863894.1 7 - 8 1312 65
XM_017008406.1 XP_016863895.1 7 - 8 1124 65
XM_017008407.1 XP_016863896.1 7 - 8 1312 65
GenBank mRNA AK001755.1 - 7 - 8 1064 65
AK027047.1 - 2 - 3 225 65
AK027413.1 - 7 - 8 1039 65
AL833208.1 - 6 - 7 910 65
BC094751.1 - 7 - 8 1278 65

View Full Product Details