The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138805.2 | NP_620160.1 | 7 - 8 | 665 | 136 |
XM_005264862.4 | XP_005264919.1 | 7 - 8 | 638 | 136 | |
XM_005264863.4 | XP_005264920.1 | 7 - 8 | 671 | 136 | |
XM_005264864.4 | XP_005264921.1 | 7 - 8 | 643 | 136 | |
XM_006712965.3 | XP_006713028.1 | 7 - 8 | 435 | 136 | |
XM_006712967.3 | XP_006713030.1 | 6 - 7 | 562 | 136 | |
XM_011533348.2 | XP_011531650.1 | 7 - 8 | 692 | 136 | |
XM_011533349.2 | XP_011531651.1 | 7 - 8 | 621 | 136 | |
XM_011533350.2 | XP_011531652.1 | 7 - 8 | 689 | 136 | |
XM_011533351.2 | XP_011531653.1 | 7 - 8 | 711 | 136 | |
XM_011533352.2 | XP_011531654.1 | 6 - 7 | 565 | 136 | |
XM_011533353.2 | XP_011531655.1 | 6 - 7 | 495 | 136 | |
XM_011533354.2 | XP_011531656.1 | 6 - 7 | 492 | 136 | |
GenBank mRNA | AB105189.1 | - | 7 - 8 | 668 | 136 |
AF494381.1 | - | 5 - 6 | 368 | 136 | |
AK127546.1 | - | 7 - 8 | 660 | 136 | |
AY358677.1 | - | 7 - 8 | 679 | 136 | |
BC015359.1 | - | 7 - 8 | 574 | 136 |