The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003659.3 | NP_003650.1 | 10 - 11 | 1147 | 126 |
XM_011512041.2 | XP_011510343.1 | 10 - 11 | 929 | 126 | |
XM_011512042.1 | XP_011510344.1 | 10 - 11 | 1096 | 126 | |
XM_011512043.2 | XP_011510345.1 | 6 - 7 | 1733 | 126 | |
GenBank mRNA | AK296239.1 | - | 10 - 11 | 927 | 126 |
AK309714.1 | - | 10 - 11 | 1246 | 126 | |
AK314259.1 | - | 10 - 11 | 1117 | 126 | |
AK316240.1 | - | 10 - 11 | 126 | ||
AY544121.1 | - | 10 - 11 | 1100 | 126 | |
BC141820.1 | - | 10 - 11 | 1112 | 126 | |
Y09443.1 | - | 10 - 11 | 1115 | 126 |