Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGPS
Entrez Gene ID
8540
Gene Name
alkylglycerone phosphate synthase
Gene Aliases
ADAP-S, ADAS, ADHAPS, ADPS, ALDHPSY, RCDP3
Chromosome Location
Chr.2: 177392743 - 177543836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003659.3 NP_003650.1 11 - 12 1287 105
XM_011512041.2 XP_011510343.1 11 - 12 1069 105
XM_011512042.1 XP_011510344.1 11 - 12 1236 105
XM_011512043.2 XP_011510345.1 7 - 8 1873 105
GenBank mRNA AK296239.1 - 11 - 12 1067 105
AK309714.1 - 11 - 12 1386 105
AK314259.1 - 11 - 12 1257 105
AK316240.1 - 11 - 12 1068 105
AY544121.1 - 11 - 12 1240 105
BC141820.1 - 11 - 12 1252 105
Y09443.1 - 11 - 12 1255 105

View Full Product Details