The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003659.3 | NP_003650.1 | 4 - 5 | 610 | 82 |
XM_011512041.2 | XP_011510343.1 | 4 - 5 | 392 | 82 | |
XM_011512042.1 | XP_011510344.1 | 4 - 5 | 559 | 82 | |
GenBank mRNA | AK296239.1 | - | 4 - 5 | 390 | 82 |
AK309714.1 | - | 4 - 5 | 709 | 82 | |
AK314259.1 | - | 4 - 5 | 580 | 82 | |
AK316240.1 | - | 4 - 5 | 391 | 82 | |
AY544121.1 | - | 4 - 5 | 563 | 82 | |
BC141820.1 | - | 4 - 5 | 575 | 82 | |
Y09443.1 | - | 4 - 5 | 578 | 82 |