The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003659.3 | NP_003650.1 | 9 - 10 | 1040 | 112 |
XM_011512041.2 | XP_011510343.1 | 9 - 10 | 822 | 112 | |
XM_011512042.1 | XP_011510344.1 | 9 - 10 | 989 | 112 | |
XM_011512043.2 | XP_011510345.1 | 5 - 6 | 1626 | 112 | |
GenBank mRNA | AK296239.1 | - | 9 - 10 | 820 | 112 |
AK309714.1 | - | 9 - 10 | 1139 | 112 | |
AK314259.1 | - | 9 - 10 | 1010 | 112 | |
AK316240.1 | - | 9 - 10 | 821 | 112 | |
AY544121.1 | - | 9 - 10 | 993 | 112 | |
BC141820.1 | - | 9 - 10 | 1005 | 112 | |
Y09443.1 | - | 9 - 10 | 1008 | 112 |