The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014633.4 | NP_055448.1 | 1 - 2 | 478 | 95 |
GenBank mRNA | AK291794.1 | - | 1 - 2 | 214 | 95 |
BC058914.1 | - | 1 - 2 | 225 | 95 | |
BP250198.1 | - | 1 - 2 | 478 | 95 | |
D63875.1 | - | 1 - 2 | 138 | 95 |