The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318766.1 | NP_001305695.1 | 2 - 3 | 262 | 82 |
NM_033388.1 | NP_203746.1 | 2 - 3 | 262 | 82 | |
XM_005274376.4 | XP_005274433.1 | 2 - 3 | 261 | 82 | |
XM_006718732.2 | XP_006718795.1 | 2 - 3 | 261 | 82 | |
XM_006718733.3 | XP_006718796.1 | 2 - 3 | 261 | 82 | |
XM_011545332.1 | XP_011543634.1 | 2 - 3 | 261 | 82 | |
XM_011545333.1 | XP_011543635.1 | 2 - 3 | 241 | 82 | |
XM_011545334.1 | XP_011543636.1 | 2 - 3 | 123 | 82 | |
GenBank mRNA | AK090597.1 | - | 2 - 3 | 253 | 82 |
AK091151.1 | - | 2 - 3 | 241 | 82 | |
AK093017.1 | - | 2 - 3 | 235 | 82 | |
AK303270.1 | - | 2 - 3 | 243 | 82 | |
BC108672.1 | - | 2 - 3 | 220 | 82 | |
BC137489.1 | - | 2 - 3 | 360 | 82 | |
BC137490.1 | - | 2 - 3 | 360 | 82 | |
BC142718.1 | - | 2 - 3 | 249 | 82 | |
BC146660.1 | - | 2 - 3 | 249 | 82 | |
BQ929430.1 | - | 2 - 3 | 273 | 82 |