The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318766.1 | NP_001305695.1 | 5 - 6 | 807 | 75 |
NM_033388.1 | NP_203746.1 | 5 - 6 | 687 | 75 | |
XM_005274376.4 | XP_005274433.1 | 5 - 6 | 686 | 75 | |
XM_006718732.2 | XP_006718795.1 | 5 - 6 | 686 | 75 | |
XM_006718733.3 | XP_006718796.1 | 5 - 6 | 686 | 75 | |
XM_011545332.1 | XP_011543634.1 | 5 - 6 | 686 | 75 | |
XM_011545333.1 | XP_011543635.1 | 5 - 6 | 1135 | 75 | |
XM_011545334.1 | XP_011543636.1 | 5 - 6 | 548 | 75 | |
GenBank mRNA | AK024423.1 | - | 1 - 2 | 411 | 75 |
AK090597.1 | - | 5 - 6 | 798 | 75 | |
AK091151.1 | - | 5 - 6 | 1135 | 75 | |
AK093017.1 | - | 5 - 6 | 660 | 75 | |
BC036713.2 | - | 3 - 4 | 416 | 75 | |
BC137489.1 | - | 5 - 6 | 785 | 75 | |
BC137490.1 | - | 5 - 6 | 785 | 75 | |
BC142718.1 | - | 5 - 6 | 674 | 75 | |
BC146660.1 | - | 5 - 6 | 674 | 75 |