The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012341.2 | NP_036473.2 | 10 - 11 | 1176 | 71 |
GenBank mRNA | AF120334.1 | - | 10 - 11 | 1128 | 71 |
AF325353.1 | - | 10 - 11 | 1105 | 71 | |
AK001548.1 | - | 10 - 11 | 1123 | 71 | |
AK001552.1 | - | 10 - 11 | 1178 | 71 | |
AK097093.1 | - | 8 - 9 | 1984 | 71 | |
AK222861.1 | - | 10 - 11 | 1123 | 71 | |
AK295231.1 | - | 9 - 10 | 1019 | 71 | |
AK301721.1 | - | 10 - 11 | 1291 | 71 | |
AK302219.1 | - | 9 - 10 | 981 | 71 | |
BC033784.1 | - | 10 - 11 | 1099 | 71 | |
BC038975.2 | - | 10 - 11 | 1137 | 71 |