Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GTPBP4
Entrez Gene ID
23560
Gene Name
GTP binding protein 4
Gene Aliases
CRFG, NGB, NOG1
Chromosome Location
Chr.10: 988409 - 1017768 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012341.2 NP_036473.2 11 - 12 1257 72
GenBank mRNA AF120334.1 - 11 - 12 1209 72
AF325353.1 - 11 - 12 1186 72
AK001548.1 - 11 - 12 1204 72
AK001552.1 - 11 - 12 1259 72
AK097093.1 - 9 - 10 2065 72
AK295231.1 - 10 - 11 1100 72
AK301721.1 - 11 - 12 1372 72
AK302219.1 - 10 - 11 1062 72
BC033784.1 - 11 - 12 1180 72
BC038975.2 - 11 - 12 1218 72

View Full Product Details