The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012341.2 | NP_036473.2 | 3 - 4 | 389 | 95 |
GenBank mRNA | AF120334.1 | - | 3 - 4 | 341 | 95 |
AF325353.1 | - | 3 - 4 | 318 | 95 | |
AK001548.1 | - | 3 - 4 | 336 | 95 | |
AK001552.1 | - | 3 - 4 | 391 | 95 | |
AK222861.1 | - | 3 - 4 | 336 | 95 | |
AK301721.1 | - | 3 - 4 | 504 | 95 | |
BC033784.1 | - | 3 - 4 | 312 | 95 | |
BC038975.2 | - | 3 - 4 | 350 | 95 |