Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTPBP4
Entrez Gene ID
23560
Gene Name
GTP binding protein 4
Gene Aliases
CRFG, NGB, NOG1
Chromosome Location
Chr.10: 988409 - 1017768 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012341.2 NP_036473.2 5 - 6 636 94
GenBank mRNA AF120334.1 - 5 - 6 588 94
AF325353.1 - 5 - 6 565 94
AK001548.1 - 5 - 6 583 94
AK001552.1 - 5 - 6 638 94
AK097093.1 - 3 - 4 1444 94
AK222861.1 - 5 - 6 583 94
AK295231.1 - 4 - 5 479 94
AK301721.1 - 5 - 6 751 94
AK302219.1 - 4 - 5 441 94
BC033784.1 - 5 - 6 559 94
BC038975.2 - 5 - 6 597 94

View Full Product Details